Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 2 µm
Leverandør: Avantor
EXL-1011-1002UEA 10425 NOK
EXL-1011-1002U EXL-1011-1003U EXL-1011-1046U EXL-1011-1202U EXL-1011-1203U EXL-1011-1246U EXL-1011-1502U EXL-1011-1503U EXL-1011-1546U EXL-1011-0202U EXL-1011-0203U EXL-1011-0302U EXL-1011-0303U EXL-1011-0346U EXL-1011-0502U EXL-1011-0503U EXL-1011-0546U EXL-1011-7502U EXL-1011-7503U EXL-1011-7546U ACE-1P11-GD4U
Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 2 µm
Kromatografikolonner
Avantor® ACE® Excel® SuperC18 columns provide high stability across an extended pH range. These stainless steel columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.
  • Ultra-inert for maximum performance and reproducibility
  • Ultra low bleed for improved LC/MS compatibility
  • Wide range of particle sizes (1,7 to 10 µm) and complementary phases
Maximum recommended pH: 1,5 to 11,5
Max. pressure: 1000 bar
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Karbonbelastning 14,8% 14,8%
Kappet ved endene Ja Yes
Maks. trykk 1000 bar
pH område 1,5 - 11,5 1,5 - 11,5
Overflate areal 400 m²/g 400 m²/g
USP kode L1 L1