Siliconising Fluid, hydrocarbon soluble

Leverandør: Thermo Fisher Scientific
Danger

TS-42800 TS-42801
HYPETS-42800EA 4425 NOK
HYPETS-42800 HYPETS-42801
Siliconising Fluid, hydrocarbon soluble
Siliconising Fluid

Thermo Scientific Hydrocarbon Soluble Siliconizing Fluid is a short chain, clear polymeric silicone fluid consisting primarily of dichlorooctamethyltetrasiloxane. When applied to glass, quartz or similar products, the unhydrolyzed chlorines present on the chain react with surface silanols to form a neutral, hydrophobic and tightly bonded film over the entire surface


  • Components: 4-Hydroxy-4-methylpentan-2-on, 2-Methyl-2-Propanol, Silan, Trimethoxyoctadecyl
  • Concentrations: 25 – 45, 25 – 45, 10 – 20 %


Our Hydrocarbon Soluble Siliconizing Fluid is ideal for use on metals, certain plastics, ceramics and fiber optics..


Our Hydrocarbon Soluble Siliconizing Fluid is acidic and care should be taken to avoid corrosion of metal that comes into contact with the liquid. The fluid is acidic only during application. After application the surface is neutral.


NB: Material is flammable before film is formed and HCI fumes are generated in the application.

CAS-nummer: 2474-02-4
FN: 2987
ADR: 8 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR