Digital scaler-timer
Leverandør: EDUCATIONSCIENCE COLLECTION P
EDSC281-006EA 0 NOK
EDSC281-006
Digital scaler-timer
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
General purpose scaler-timer, suitable for radioactivity counting utilising a GM tube and as a general purpose millisecond digital timing device with manual or electronic triggering by a photo gate.
  • 6-digit, bright green LED display
  • Capacity: 999,999 counts or seconds
  • Manual or timed counting of 1,10 or 100 seconds
  • Press-to-read switch to read the accurate applied voltage to the GM tube
  • Auto ranging on timing modes
  • Built in loudspeaker and switch on the back of the machine allows audible indication of counts when required.