Spectral analysis kit
Leverandør: EDUCATIONSCIENCE COLLECTION P
EDSC3565EA 0 NOK
EDSC3565
Spectral analysis kit
Lærings materiell Læremateriale for fysikk Optics Learning Activities
Students observe the characteristic colour imparted to a flame from various elements. They also observe that different distinguishing colors. Using the spectroscope they see the light emitted from familiar light sources and compare the appearances.
Leveringsinformasjon: Kit includes 24 spectroscopes 15 handles one length of wire (1,22 m) and one bottle each of chloride, potassium chloride, potassium nitrate, sodium, hydrogen carbonate, sodium chloride, strontium chloride and strontium nitrate.