Disposable samplers, ZoneDispo
Leverandør: BURKLE
5393-4420 5393-4430 5393-4421 5393-4431
300-0212EA 6375 NOK
300-0212 300-0243 300-0213 300-0244
Disposable samplers, ZoneDispo
Prøvetakere Prøvetakere til engangsbruk
Disposable ZoneDispo samplers for pourable bulk goods up to 2 mm particle size.
  • Three openings of Ø 11 mm