Single gas detectors, BW Clip
Leverandør: BW TECHNOLOGIES
BWC2-H510 BWC2-M3060 BWC2-S25 BWC2-X BWC2-M30200 BWC2-H BWC2-M BWC2R-X
151-0229EA 2650 NOK
151-0229 118-0484 151-0232 151-0231 118-0483 151-0228 151-0230 151-0322
Single gas detectors, BW Clip
Gassdeteksjonssystemer
Single gas detectors for safety and compliance in harsh environments with extreme temperatures.
  • Maintenance-free: No sensor or battery change
  • Hibernation mode with case accessory or IntelliDox
  • Automated self-test of battery, sensor and electronics
  • Wide-angle flash, which alerts with audible and vibrating alarm
  • Automatic logging of the 35 latest events and bump test results
Sertifiseringer: ATEX: CE 0539 II 1G; Ex ia IIC T4 Ga; DEMKO 14 ATEX 1356 - IECEx: Ex ia IIC T4 Ga; IECEx UL 14.0063
Leveringsinformasjon: Supplied with specified sensor,More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Fuktighetsområde (RF) 5 - 95 %
Alarm Visual, vibrating, audible
levetid batteri 2 years
B×D×H 41×50×87 mm
Vekt 92 g