Cell Viability Reagent Kits for Beckman Coulter Vi-CELL® systems
Leverandør: CELL VIABILITY
2400101 2400102
2400101.EA 15100 NOK
2400101. 2400102.
Cell Viability Reagent Kits for Beckman Coulter Vi-CELL® systems
Assay Cellulær analyse
The Cell Viability Reagent Kits are designed specifically for use with the Beckman Coulter Vi-CELL® Cell Viability Analyser. The Cell Viability Reagent Kits, as well as being cost-efficient, are proven to be of equivalent quality and performance when compared to Beckman Coulter Vi-CELL® Kits.
The Vi-CELL® Cell Viability Analyser is an automated instrument that performs the trypan blue dye exclusion method for cellular viability determination. This data is important for the monitoring of basic tissue and cell culture conditions in bioreactors. Cell and tissue culture applications in life science research, including recombinant protein and biopharmaceutical production, require cell viability information constantly. The Vi-CELL® automates the widely accepted trypan blue cell exclusion method which has been historically done using a light microscope and a haemacytometer.
Vi-CELL® is a registered trademark of Beckman Coulter Inc.
Leveringsinformasjon: Cell Viability Reagent Single Kit (equivalent to Beckman Coulter Vi-CELL® Single Pak, part number 383260) includes one reagent pack that fits inside the Vi-CELL® instrument and one bag with 200 sample cups. Cell Viability Reagent Quad Kit (equivalent to Beckman Coulter Vi-CELL® Quad Pak, part number 383722) includes four reagent packs that fit inside the Vi-CELL® instrument and 4×200 sample cups.