Hybrid multi-parameter meter, handheld, SL1000 PPA
Leverandør: Hach
LPV4439910002 LPV4439920002
705-0941EA 53855.2 NOK
705-0941 705-0942
Hybrid multi-parameter meter, handheld, SL1000 PPA
Elektrokjemiske målere pH-meter
The SL1000 is a portable colorimeter that provides 75% faster testing by using Chemkey® reagents instead of powder pillows or cuvette test methods. No reagent handling is necessary − all chemical and processes are entirely contained inside the Chemkey®. Besides that, the meter can be used in combination with digital IntelliCAL™ electrodes to measure different electrochemical parameters, such as pH, conductivity, ORP, dissolved oxygen, nitrate, fluoride in clean water applications.
  • Ideal for disinfection control in the field or clean water treatment
More Product Information
Bestill nå
SPESIFIKASJONER
Modell SL1000
Environmentally Preferable
temperaturkompensering Off, automatic and manual (for IntelliCal™ probes; parameter dependent)
Strømforsyning Lithium ion rechargeable battery or external charger (100 - 240 V, 50/60 Hz)
Samsvar IP 64 with caps installed for the meter probe connector ports or with the probes connected to the instrument.
Datalogg - loggfunksjon Automatic in Press to Read measurement mode; manual in Continuous measurement mode
Datalogg kapasitet 1000 measured values (result, date, time, site ID, user ID)
Tilkoblinger 2× M12 connectors for IntelliCal™ probes, 4× Chemkey slots, 2,5 mm Jack for external power supply
Outputs Mini USB
W×D×H 131×59×258 mm
Weight 1,2 kg