Industrial gloves, uvex phynomic XG
Supplier: UVEX
6007009 6007007 6007008 6007010 6007011 6007006 6007012 6007005
112-4425EA 1050 NOK
112-4425 112-4423 112-4424 112-4426 112-4427 112-4523 UVEX6007012 UVEX6007005
Industrial gloves, uvex phynomic XG
Gloves
The uvex phynomic XG offers the best oil grip in its class. The polyamide/elastane liners feature an innovative aqua-polymer Xtra Grip foam coating which is also particularly flexible and extremely robust.
  • Ultra-slow liquid breakthrough provides additional skin protection
  • High abrasion resistance, high levels of dexterity
  • Second skin fit gives a natural touch
  • Flexible construction reduces fatigue
  • Free from allergenic substances - no harmful substances used in the manufacturing
  • Suitable for the most allergy-sensitive wearers
Ideal for fine assemblyMore Product Information
New Product
Order Now
SPECIFICATIONS
Model XG
Environmentally Preferable
Ambidextrous / hand-specific Hand-specific
Disposable / reusable Reusable
Liner material Polyamide/elastane
Finish Aqua polymer Xtra Grip foam coating on palm and fingertips
Cuff style Knit wrist
EN norm EN 388:2016 (4121X)