Membranfiltre, type 111
Leverandør: Sartorius
SART11104-293-NEA 29900 NOK
SART11104-293-N 611-0673 611-0670 SART11106-110-N SART11107-100-N 512-4863 512-4718 SART11106-293-N SART11104-142-N 611-0668 512-4872 512-4114 512-4873 SART11106-30-N 512-4550 512-4218 512-4416 512-4714 512-4516 512-4318 512-4913 512-4715 512-4418 512-4616 512-4518 512-4716 512-4914 512-4214 512-4314 512-4116 512-4215 512-4216 512-4315 512-4414 512-4415 512-4316 512-4118 SART11107-293-N SART11107-30-N
Membranfiltre, type 111
Filter Membranfilter
Cellulose acetate hydrophilic membrane filters. Ideal for biological and clinical analysis, sterility tests and scintillation measurements. Very low protein binding capacity, suitable for aqueous, alcoholic media and hot gases.
* Sterile