Zirconium oxide polymeric precursor
Leverandør: Thermo Scientific
|
Danger
|
42766.14
42766.22
42766.14EA
2575
NOK
42766.14
42766.22
Zirconium oxide polymeric precursor
Zirconium oxide
Physical Form: Viscous liquid
Sensitivity: Moisture Sensitive
Appearance: Yellow to brown
Odour: Alcohol-like
Solubility: Soluble in alcohols
UN No.: UN1993
MDL No.: MFCD00011310
Applications: Preparation of zirconium oxide films, solid electrolyte
Formel:
OZr MW: 107,22 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00011310 CAS-nummer: 12036-01-0 FN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...