Metylenblått 1% (w/v) vandig løsning
Leverandør: Thermo Scientific
Synonymer: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
42771.AE 42771.AP
42771.AEEA 485 NOK
42771.AE 42771.AP
Metylenblått 1% (w/v) vandig løsning
Metylenblått
Molecular Formula: C16H18ClN3S Formula Weight: 319.86 Physical Form: Liquid MDL No.: MFCD00012111
Formel: C₁₆H₁₈ClN₃S
MW: 319,86 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150008
CAS-nummer: 61-73-4