Tin(II) pyrophosphate
Leverandør: Thermo Scientific
|
Warning
|
89785.30
89785.18
89785.30EA
2250
NOK
89785.30
89785.18
Tin(II) pyrophosphate
Tin(II) pyrophosphate
CAS No.: 15578-26-4
Molecular Formula: Sn2P2O7
Formula Weight: 411.32
Physical Form: Powder
Density: 4.009
Melting Point: 400° dec.
Solubility: Insoluble in water
MDL No.: MFCD00049544
Formel:
O₇P₂Sn₂ MW: 411,361 g/mol Smeltepunkt: 400 °C (dec.) Tetthet: 4,009 Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00049544 CAS-nummer: 15578-26-4 EINECS: 239-635-5 Merck Indeks: 14,8788 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...