3-Ethoxy-2-cyclohexen-1-one 99%
Leverandør: Thermo Scientific
A14224.09
A14224.18
A14224.30
A14224.09EA
700
NOK
A14224.09
A14224.18
A14224.30
3-Ethoxy-2-cyclohexen-1-one 99%
3-Ethoxy-2-cyclohexen-1-one
Purity: 99%
CAS No.: 5323-87-5
Molecular Formula: C8H12O2
Formula Weight: 140.18
Storage Temperature: -30°C to -10°C
Flash Point: 107°(224°F)
Refractive Index: 1.5060
Boiling Point: 76-78°/1mm
Beilstein Reference: 2042049
MDL No.: MFCD00001580
Formel:
C₈H₁₂O₂ MW: 140,18 g/mol Kokepunkt: 76…78 °C (1 mmHg) Tetthet: 1,04 Flash punkt: 107 °C (224 °F) |
MDL nummer:
MFCD00001580 CAS-nummer: 5323-87-5 EINECS: 226-190-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...