DL-Histidine monohydrochloride monohydrate 99%
Leverandør: Thermo Scientific
Synonymer: H-DL-His-OH.HCl.H2O, H-DL-His-OH·HCl·H₂O
A16165.06 A16165.14 A16165.22
A16165.06EA 485 NOK
A16165.06 A16165.14 A16165.22
DL-Histidine monohydrochloride monohydrate 99%
DL-Histidine monohydrochloride monohydrate
Purity: 99% CAS No.: 123333-71-1 Molecular Formula: C6H9N3O2.HCl.H2O Formula Weight: 209.63 (191.62anhy) Melting Point: ca 250° dec. MDL No.: MFCD00151028
Formel: C₆H₉N₃O₂·HCl·H₂O
MW: 209,63 g/mol (191,62 anhy)
Smeltepunkt: ∼ 250 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00151028
CAS-nummer: 123333-71-1