2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%
Leverandør: Thermo Scientific
Warning
Synonymer: racemic-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, rac-BINAP, (+/-)-(1,1'-Binaphthalene-2,2'-diyl)bis(diphenylphosphine), (±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (+/-)-BINAP
H26226.06 H26226.14
H26226.06EA 2850 NOK
H26226.06 H26226.14
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Purity: 98% CAS No.: 98327-87-8 Molecular Formula: C44H32P2 Formula Weight: 622.69 Melting Point: 281-285° Beilstein Reference: 5321443 MDL No.: MFCD00010805
Formel: C₄₄H₃₂P₂
MW: 622,67 g/mol
Smeltepunkt: 281…285 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010805
CAS-nummer: 98327-87-8
EINECS: None
Merck Indeks: 14,1223