Trimethyl(vinyloxy)silane 97%
Leverandør: Thermo Scientific
|
Danger
|
H26876.03
H26876.06
H26876.03EA
455
NOK
H26876.03
H26876.06
Trimethyl(vinyloxy)silane 97%
Trimethyl(vinyloxy)silane
Purity: 97%
CAS No.: 6213-94-1
Molecular Formula: H2C=CHOSi(CH3)3
Formula Weight: 116.24
Flash Point: -19°(-2°F)
Refractive Index: 1.389
Boiling Point: 74°
UN No.: UN1993
MDL No.: MFCD00054764
Formel:
C₅H₁₂OSi MW: 116,24 g/mol Kokepunkt: 74 °C Tetthet: 0,779 Flash punkt: –19 °C (–2 °F) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00054764 CAS-nummer: 6213-94-1 EINECS: 228-281-7 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...