Ethyl-3-cyano-3-phenylpyruvate 97%
Leverandør: Thermo Scientific
Warning
H26951.06 H26951.14
H26951.06EA 620 NOK
H26951.06 H26951.14
Ethyl-3-cyano-3-phenylpyruvate 97%
Ethyl-3-cyano-3-phenylpyruvate
Purity: 97% CAS No.: 6362-63-6 Molecular Formula: C12H11NO3 Formula Weight: 217.23 Melting Point: 125-129° MDL No.: MFCD00019801
Formel: C₁₂H₁₁NO₃
MW: 217,23 g/mol
Smeltepunkt: 125…129 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00019801
CAS-nummer: 6362-63-6
EINECS: 228-844-7