6-Bromopyrazolo[1,5-a]pyrimidine 97%
Leverandør: Thermo Scientific
Warning
H33855.03 H33855.06
H33855.03EA 1250 NOK
H33855.03 H33855.06
6-Bromopyrazolo[1,5-a]pyrimidine 97%
6-Bromopyrazolo[1,5-a]pyrimidine
Purity: 97% CAS No.: 705263-10-1 Molecular Formula: C6H4BrN3 Formula Weight: 198.02 Melting Point: 122-124° MDL No.: MFCD09832894
Formel: C₆H₄BrN₃
MW: 198,02 g/mol
Smeltepunkt: 122…124 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09832894
CAS-nummer: 705263-10-1