Agmatine sulphate 97% (by HPLC)
Leverandør: Thermo Scientific
Synonymer: 1-Amino-4-guanidobutane Sulphate, 1-(4-Aminobutyl)guanidine sulphate
H55363.MD H55363.03 H55363.06
H55363.MDEA 575 NOK
H55363.MD H55363.03 H55363.06
Agmatine sulphate 97% (by HPLC)
Agmatine sulphate
Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine.
Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.
Formel: C₅H₁₄N₄
MW: 228,27 g/mol
Smeltepunkt: 231…238 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013109
CAS-nummer: 2482-00-0
EINECS: 219-617-3
Merck Indeks: 14,188
Bestill nå
Spesifikasjon Testresultater
Assay (HPLC) >96.0%