2-chloro-5-iodo-3-(trifluoromethyl)pyridine 95%
Leverandør: Thermo Scientific
|
Warning
|
Synonymer:
2-Chloro-5-iodo-3-(trifluoromethyl)pyridine
H64329.03
H64329.06
H64329.03EA
910
NOK
H64329.03
H64329.06
2-chloro-5-iodo-3-(trifluoromethyl)pyridine 95%
2-chloro-5-iodo-3-(trifluoromethyl)pyridine
Purity: 95%
CAS No.: 887707-25-7
Molecular Formula: C6H2ClF3IN
Formula Weight: 307.44
Physical Form: Solid
Sensitivity: Light Sensitive
Melting Point: 43°
Beilstein Reference: 10249610
MDL No.: MFCD12024302
Formel:
C₆H₂ClF₃IN MW: 307,439 g/mol Smeltepunkt: 43 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD12024302 CAS-nummer: 887707-25-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...