4-Bromo-1H-pyrazole 98+%
Leverandør: Thermo Scientific
|
Warning
|
H64673.14
H64673.36
H64673.14EA
905
NOK
H64673.14
H64673.36
4-Bromo-1H-pyrazole 98+%
4-Bromo-1H-pyrazole
Purity: 98+%
CAS No.: 2075-45-8
Molecular Formula: C3H3BrN2
Formula Weight: 146.97
Physical Form: Solid
Melting Point: 93-96° lit.
Boiling Point: 250-260° lit.
Beilstein Reference: 106510
MDL No.: MFCD00075602
Formel:
C₃H₃BrN₂ MW: 146,97 g/mol Kokepunkt: 250…260 °C (lit.) Smeltepunkt: 93…96 °C (lit.) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00075602 CAS-nummer: 2075-45-8 |