Ethyl-2,4-dichloropyrimidine-5-carboxylate 98%
Leverandør: Thermo Scientific
|
Warning
|
Synonymer:
Ethyl 2,4-Dichloro-5-pyrimidinecarboxylate
H64788.03
H64788.06
H64788.03EA
1625
NOK
H64788.03
H64788.06
Ethyl-2,4-dichloropyrimidine-5-carboxylate 98%
Ethyl-2,4-dichloropyrimidine-5-carboxylate
Purity: 98%
CAS No.: 51940-64-8
Molecular Formula: C7H6Cl2N2O2
Formula Weight: 221.04
Physical Form: Powder
Appearance: White
Melting Point: 36-37°
Beilstein Reference: 152173
MDL No.: MFCD09910281
Formel:
C₇H₆Cl₂N₂O₂ MW: 221,04 g/mol Smeltepunkt: 36…37 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD09910281 CAS-nummer: 51940-64-8 EINECS: 257-531-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...