Adenosine-5'-diphosphate monopotassium salt dihydrate 99%
Leverandør: Thermo Scientific
Synonymer:
5'-ADP-K dihydrate, Potassium (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-4-hydroxy-5-(((hydroxy(phosphonooxy)phosphoryl)oxy)methyl)tetrahydrofuran-3-olate dihydrate, ADP-K dihydrate
Purity: 99%
CAS No.: 72696-48-1
Molecular Formula: C10H14KN5O10P2.2H2O
Formula Weight: 501.33 (465.29anhy)
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline powder
Appearance: White
MDL No.: MFCD00066472
Formel:
C₁₀H₁₄KN₅O₁₀P₂·2H₂O MW: 501,33 g/mol |
MDL nummer:
MFCD00066472 CAS-nummer: 72696-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...