Praziquantel 98%

Leverandør: Thermo Scientific
Warning

J66188.06 J66188.14
J66188.06EA 1225 NOK
J66188.06 J66188.14
Praziquantel 98%
Praziquantel

Purity: 98% CAS No.: 55268-74-1 Molecular Formula: C19H24N2O2 Formula Weight: 312.41 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: White to off-white Melting Point: 136-138° Boiling Point: 1377° Solubility: Freely soluble in ethanol or dichloromethane. Slightly soluble in water Beilstein Reference: 761557 MDL No.: MFCD00058531


Applications: An anti-helminthic compound

Formel: C₁₉H₂₄N₂O₂
MW: 312,41 g/mol
Kokepunkt: 1377 °C
Smeltepunkt: 136…138 °C
Flash punkt: >110 °C (230 °F)
MDL nummer: MFCD00058531
CAS-nummer: 55268-74-1
EINECS: 259-559-6
Merck Indeks: 14,7716

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR