2,2'-Bipyrimidyl 96%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: 2,2'-Dipyrimidyl, 2,2'-Bipyrimidine

L01190.03 L01190.MD
L01190.03EA 4125 NOK
L01190.03 L01190.MD
2,2'-Bipyrimidyl 96%
2,2'-Bipyrimidyl

Purity: 96% CAS No.: 34671-83-5 Molecular Formula: C8H6N4 Formula Weight: 158.16 Melting Point: 110-116° Beilstein Reference: 607396 MDL No.: MFCD00014600

Formel: C₈H₆N₄
MW: 158.16 g/mol
Smeltepunkt: 110…116 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00014600
CAS-nummer: 34671-83-5
EINECS: 252-137-2

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR