2-(2-Thienyl)pyridine 97%

Leverandør: Thermo Scientific
Warning

Synonymer: 2-(Thiophen-2-yl)pyridine, 2-(2-Pyridyl)thiophene

L04775.03 L04775.06
L04775.03EA 840 NOK
L04775.03 L04775.06
2-(2-Thienyl)pyridine 97%
2-(2-Thienyl)pyridine

Purity: 97% CAS No.: 3319-99-1 Molecular Formula: C9H7NS Formula Weight: 161.23 Melting Point: 60-65° Beilstein Reference: 508454 MDL No.: MFCD00044441

Formel: C₉H₇NS
MW: 161,22 g/mol
Smeltepunkt: 60…65 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00044441
CAS-nummer: 3319-99-1
EINECS: 222-022-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR