Methyl-2,2-dimethylacetoacetate 99%

Leverandør: Thermo Fisher Scientific

Synonymer: Methyl 2,2-dimethylacetoacetate

L10917.03 L10917.06
L10917.03EA 615 NOK
L10917.03 L10917.06
Methyl-2,2-dimethylacetoacetate 99%
Methyl-2,2-dimethylacetoacetate

Purity: 99% CAS No.: 38923-57-8 Molecular Formula: C7H12O3 Formula Weight: 144.17 Refractive Index: 1.4205 Boiling Point: 72-74°/15mm Beilstein Reference: 1753940 MDL No.: MFCD00026232

Formel: C₇H₁₂O₃
MW: 144.17 g/mol
Kokepunkt: 72…74 °C (15 mmHg)
Tetthet: 0.999
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00026232
CAS-nummer: 38923-57-8
EINECS: 254-189-1

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR