Cyclohexyl acrylate ≥98% stabilised

Leverandør: Thermo Scientific
Warning


Stabilisering: Stabilised with 4-methoxyphenol (MEHQ) 100 ppm

L12925.14 L12925.06
L12925.14EA 1050 NOK
L12925.14 L12925.06
Cyclohexyl acrylate ≥98% stabilised
Cyclohexyl acrylate

Stab. with 100ppm 4-methoxyphenol


Purity: 98+% CAS No.: 3066-71-5 Molecular Formula: C9H14O2 Formula Weight: 154.21 Storage Temperature: 2°C to 8°C Flash Point: 68°(154°F) Refractive Index: 1.4590 Boiling Point: 182-184° Beilstein Reference: 1937528 UN No.: UN3082 MDL No.: MFCD00046349

Formel: C₉H₁₄O₂
Kokepunkt: 182…184 °C
Tetthet: 0,980
Flash punkt: 68 °C (154 °F)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00046349
CAS-nummer: 3066-71-5
EINECS: 221-319-3
FN: 3082
ADR: 9,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR