2-(Hydroxymethyl)-2-nitro-1,3-propanediol 95%
Leverandør: Thermo Scientific
Warning
Synonymer: 2-(Hydroxymethyl)-2-nitropropane-1,3-diol, Tris(hydroxymethyl)nitromethane, Trimethylolnitromethane
L13349.22 L13349.36
L13349.22EA 485 NOK
L13349.22 L13349.36
2-(Hydroxymethyl)-2-nitro-1,3-propanediol 95%
2-(Hydroxymethyl)-2-nitro-1,3-propanediol
Purity: 95% CAS No.: 126-11-4 Molecular Formula: C4H9NO5 Formula Weight: 151.12 Melting Point: ca 160° dec. Beilstein Reference: 742427 MDL No.: MFCD00007395
Formel: C₄H₉NO₅
MW: 151,12 g/mol
Smeltepunkt: ∼ 160 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007395
CAS-nummer: 126-11-4
EINECS: 204-769-5
Merck Indeks: 14,9753