Laboratory thermometers, conformity certified, TFX 422C
Leverandør: EBRO
1340-5433 1340-5434
EBRO1340-5433EA 9250 NOK
EBRO1340-5433 EBRO1340-5434
Laboratory thermometers, conformity certified, TFX 422C
Termometere Digitaltermometer
These ultra accurate thermometers are suitable for high-precision temperature monitoring in food applications and in the laboratory.
  • Robust and impact resistant
  • Functions: MIN/MAX and hold
  • High accuracy
  • Waterproof (IP 67)
  • Available with 60 cm or 150 cm silicone cable
With fixed PT1000 probe.
Sertifiseringer: DIN EN 13485. Conformity certified; certified with the EB 4401 food inspection case.