Bariumklorid vandig løsning USP Test Solution (TS)
Leverandør: VWR Chemicals
85332.180EA 530 NOK
85332.180 85332.260
Bariumklorid vandig løsning USP Test Solution (TS)
Bariumklorid
Ready to use test/reagent solutions (TS and RS).
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Formel: BaCl₂
MW: 208,24 g/mol
Smeltepunkt: ~ 963 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003445
CAS-nummer: 10361-37-2
EINECS: 0
FN: 3287
ADR: 6.1,III
Merck Indeks: 12,00998