Pierce™ Micro BCA™ Protein Assay Kit and Reagents
Leverandør: Thermo Fisher Scientific
23235
23231
23232
23234
PIER23235EA
4200
NOK
PIER23235
PIER23231
PIER23232
PIER23234
Pierce™ Micro BCA™ Protein Assay Kit and Reagents
Assay
Proteinanalyser
Micro BCA™ protein assay kit is a specialised version of Pierce™ BCA protein assay for determining the protein concentration of dilute samples.
- Colorimetric method read at 562 nm
- Linear working range for BSA is 0,5 to 20 µg/ml
- Compatible with most ionic and non ionic detergents
- Less protein variation than dye-binding methods
- All kit reagents stable at room temperature for two years
- Working reagent is stable for 24 hours
- Convenient for microplate or cuvette format
Mixing together the three Micro BCA™ reagents results in a working solution that is sufficiently concentrated to measure protein when mixed with an equal volume of sample. The result is an assay for accurately measuring 0,5 to 20 µg/ml protein solutions. The assay is exceptionally linear and exhibits very low levels of protein-to-protein variability.