Menthofuran 95%
Leverandør: Thermo Scientific
|
Warning
|
413560100
413560500
ACRO413560100EA
830
NOK
ACRO413560100
ACRO413560500
Menthofuran 95%
Menthofuran
CAS No.: 494-90-6
Formel:
C₁₀H₁₄O Kokepunkt: 87 °C (15 mmHg) Tetthet: 0,97 g/cm³ Flash punkt: 76 °C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00041851 CAS-nummer: 494-90-6 |
Spesifikasjon Testresultater
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4810 to 1.4870 (20°C, 589 nm) |
GC | ≥94 % |
Distillation range | 170 to 195 °C |
Aromatics | ≤50 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...