Methoxytrimethylsilane 96+%
Leverandør: Thermo Scientific
|
Danger
|
Synonymer:
Trimethyl(methoxy)silane
392470250
392471000
392472500
ACRO392470250EA
970
NOK
ACRO392470250
ACRO392471000
ACRO392472500
Methoxytrimethylsilane 96+%
Methoxytrimethylsilane
Formel:
C₄H₁₂OSi MW: 104,22 g/mol Kokepunkt: 49…51 °C (1013 hPa) Tetthet: 0,756 g/cm³ Flash punkt: –11 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00008344 CAS-nummer: 1825-61-2 FN: 1993 ADR: 3,II |
Spesifikasjon Testresultater
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3660 to 1.3680 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...