2,2-Dimethylcyclopentanone 97%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: 2,2-Dimethylcyclopentan-1-one

399400010 399400050
ACRO399400010EA 780 NOK
ACRO399400010 ACRO399400050
2,2-Dimethylcyclopentanone 97%
2,2-Dimethylcyclopentanone

Appearance: Clear colorless to yellow Liquid

Formel: C₇H₁₂O
MW: 112.172 g/mol
Kokepunkt: 143…145 °C (760 mmHg)
Tetthet: 0.89 g/cm³
Flash punkt: 32 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00049164
CAS-nummer: 4541-32-6
FN: 1224
ADR: 3 III

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4320 to 1.4340 (20°C, 589 nm)
GC ≥96.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR