Androsterone 97%

Leverandør: Thermo Scientific
Danger

Synonymer: Androstan-17-one, 3-hydroxy-, (3alpha,5alpha)-, 3-alpha-Hydroxy-5-alpha-androstan-17-one, 5alpha-Androsterone

204190050 204190010
ACRO204190050EA 4800 NOK
ACRO204190050 ACRO204190010
Androsterone 97%
Androsterone
Formel: C₁₉H₃₀O₂
MW: 290,447 g/mol
Smeltepunkt: 180…185 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003618
CAS-nummer: 53-41-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to light beige Powder or crystals
Infrared spectrum Conforms
GC ≥96.0 %
Specific optical rotation +90° to +98° (20°C, 589 nm) (c=0.7, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR