3-Amino-1-adamantanol 96%

Leverandør: Thermo Scientific
Warning

Synonymer: 3-Aminoadamantan-1-ol

435380010 435380050
ACRO435380010EA 810 NOK
ACRO435380010 ACRO435380050
3-Amino-1-adamantanol 96%
3-Amino-1-adamantanol
Formel: C₁₀H₁₇NO
MW: 167,25 g/mol
Smeltepunkt: 263…264 °C
Flash punkt: 110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01821204
CAS-nummer: 702-82-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Water ≤10 % (K.F.)
Elemental Analysis 7.55 to 8.35 % N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR