6-Bromonicotinic acid 96%
Leverandør: Thermo Scientific
|
Warning
|
Synonymer:
6-Bromopyridine-3-carboxylic acid, 2-Bromo-5-pyridinecarboxylic acid, 6-Bromo-3-pyridinecarboxylic acid
435400010
435400100
ACRO435400010EA
550
NOK
ACRO435400010
ACRO435400100
6-Bromonicotinic acid 96%
6-Bromonicotinic acid
Formel:
C₆H₄BrNO₂ MW: 202,01 g/mol Smeltepunkt: 179…181 °C (dec.) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD01927100 CAS-nummer: 6311-35-9 |
Spesifikasjon Testresultater
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...