3-Aminophenylboronic acid hydrochloride 98%
Leverandør: Thermo Fisher Scientific
Warning
Synonymer: (3-Aminophenyl)boronic acid hydrochloride, 3-Aminobenzeneboronic acid hydrochloride
435530010 435530050
ACRO435530010EA 1100 NOK
ACRO435530010 ACRO435530050
3-Aminophenylboronic acid hydrochloride 98%
3-Aminophenylboronic acid hydrochloride
Formel: C₆H₉BClNO₂
MW: 173.41 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00191748
CAS-nummer: 85006-23-1
Bestill nå
Spesifikasjon Testresultater
Appearance Off-white to grey Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.0 %
Elemental Analysis 7.88 to 8.28 % N