5-Acetyl-2-methoxypyridine 97%

Leverandør: Thermo Fisher Scientific
Warning

438170010 438170050
ACRO438170010EA 890 NOK
ACRO438170010 ACRO438170050
5-Acetyl-2-methoxypyridine 97%
5-Acetyl-2-methoxypyridine
Formel: C₈H₉NO₂
Smeltepunkt: 53…54 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD05664350
CAS-nummer: 213193-32-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 52.0 to 57.0 °C
Water ≤3.00 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR