(±)-Boc-α-phosphonoglycine trimethyl ester 97%

Leverandør: Thermo Scientific
Danger

Synonymer: Methyl 2-((tert-butoxycarbonyl)amino)-2-(dimethoxyphosphoryl)acetate

439210010 439210050
ACRO439210010EA 725 NOK
ACRO439210010 ACRO439210050
(±)-Boc-α-phosphonoglycine trimethyl ester 97%
(±)-Boc-α-phosphonoglycine trimethyl ester
Formel: C₁₀H₂₀NO₇P
MW: 297,24 g/mol
Smeltepunkt: 69,7…71.6 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02178893
CAS-nummer: 89524-98-1
FN: 2811
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Powder
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR