Reporter assay kits, SpectraMax® Glo Steady-Luc™
Leverandør: MOLECULAR DEVICES
R8352 R8353
MLDVR8352EA 0 NOK
MLDVR8352 MLDVR8353
Reporter assay kits, SpectraMax® Glo Steady-Luc™
Assay Cellulær analyse
The SpectraMax® Glo Steady-Luc™ Reporter Assay Kit provides a highly sensitive assay for the quantitation of firefly luciferase expression in mammalian cells. The assay kit is optimised for Molecular Devices SpectraMax® microplate readers with a preconfigured protocol provided in SoftMax® Pro Software for simplified data acquisition and analysis.
  • Glow-type luminescence provides an extended signal time window enabling batch processing of plates in screening assays
  • Optimised for luminescence microplate readers
  • Simplified data acquisition and analysis with preconfigured protocols in SoftMax® Pro Software
The application of luciferase-based reporter assay using luminescence microplate readers has become increasing popular for high-throughput analysis of gene expression as well as drug discovery applications. The SpectraMax® Glo Steady-Luc Reporter Assay Kit is designed for optimal performance on microplate readers with high throughput enabled by both the microplate format and assay signal stability.
Leveringsinformasjon: Kit includes SpectraMax® Glo Steady-Luc™ Assay Buffer, lyophilised (20 or 100 ml), Reconstitution Buffer (20 or 100 ml) and D-Luciferin (5 or 25 mg).