DNA quantitation, SpectraMax® Quant™ dsDNA assay kits
Leverandør: MOLECULAR DEVICES
R8356 R8357 R8358 R8359 R8354 R8355
MLDVR8356EA 0 NOK
MLDVR8356 MLDVR8357 MLDVR8358 MLDVR8359 MLDVR8354 MLDVR8355
DNA quantitation, SpectraMax® Quant™ dsDNA assay kits
Assay Cellulær analyse
SpectraMax® Quant™ dsDNA Assay kits are designed for fluorescence-based dsDNA quantitation across a broad range of concentrations.
  • Tailored to meet different needs
  • Highly sensitive, accurate and easy to use
  • Optimised for fluorescence microplate readers, such as SpectraMax® readers
  • Preconfigured protocols provided in SoftMax® Pro Software for simplified data acquisition and analysis
Quantitation of DNA is a critical step in molecular biology requiring accuracy, reliability, and the use of increasingly smaller sample volumes for applications such as next-generation sequencing. Compared to spectrophotometric DNA quantification, the fluorometric method provides key advantages such as significantly increased sensitivity, high selectivity for double-stranded DNA (dsDNA) over single-stranded DNA (ssDNA) or RNA, and improved contaminant tolerance (protein and carbohydrate molecules).