Antimicrobial hand cleansing dispensers, Cleanse Antibac
Leverandør: SC Johnson
Cleanse Antibac
115-2258EA 250 NOK
115-2258 115-2263 115-2264
Antimicrobial hand cleansing dispensers, Cleanse Antibac
Såpedispensere og Lotion Såpedispensere
This white antimicrobial hand cleansing dispenser is ideal for use in industry, offices, medical rooms and laboratories, food processing and manufacturing.
  • Wall-mounted
  • Durable plastic
  • One hand operation
For use with AgroBAC LOTION Wash or PureBAC FOAM Wash cartridges.