Fluid aspirator, Galtek®
Leverandør: ENTEGRIS
226-7 226-5 226-1 226-2 226-3 226-4 226-6
ENTE226-7EA 0 NOK
ENTE226-7 ENTE226-5 ENTE226-1 ENTE226-2 ENTE226-3 ENTE226-4 ENTE226-6
Fluid aspirator, Galtek®
Pumper Aspirasjonssystemer
Galtek® aspirator is ideal for the extraction and/or dilution of fluids from chemical tanks.
  • Made from PFA for long lasting corrosion resistance
  • Requires only water or air to operate
Bestill nå
SPESIFIKASJONER
Modell 226-7 226-5 226-1 226-2 226-3 226-4 226-6
Environmentally Preferable
Orifice 1,0 mm 1,8 mm 1,8 mm 1,8 mm 1,8 mm 1,8 mm 1,8 mm
Inngang/utgang 3/8” (9,53 mm) FNPT / 3/8” (9,53 mm) FNPT 3/8” (9,53 mm) FNPT / 3/8” (9,53 mm) FNPT 3/8” (9,53 mm) FNPT / 3/8” (9,53 mm) FNPT 3/8” (9,53 mm) FNPT / 1/2” (12,7 mm) O.D. tube 3/8” (9,53 mm) FNPT / 3/8” (9,53 mm) FNPT 3/8” (9,53 mm) FNPT / 1/2” (12,7 mm) O.D. tube 3/8” (9,53 mm) FNPT / 3/8” (9,53 mm) FNPT
Lengde vektstang 1/4”(6,35 mm) FNPT 1/4”(6,35 mm) FNPT 3/8” (9,53 mm) O.D. tube 1/4”(6,35 mm) FNPT 3/8” (9,53 mm) O.D. tube 1/4”(6,35 mm) FNPT 1/4”(6,35 mm) FNPT
Trykk 5−125 PSIG 5−125 PSIG 5−125 PSIG 5−125 PSIG 5−125 PSIG 5−125 PSIG 5−125 PSIG
Temperatur −18...+65 °C −18...+65 °C −18...+65 °C −18...+65 °C −18...+65 °C −18...+65 °C −18...+65 °C