Water purification system, WaterPro® BT
Leverandør: LABCONCO
9015030 9015130
LBCP9015030EA 0 NOK
LBCP9015030 LBCP9015130
Water purification system, WaterPro® BT
Vannrensesystemer Rene og ultrarene vannsystemer
The WaterPro® BT Water Purification System produces both RO and Type I water directly from tap water. The benchtop design is compact and lightweight so it can easily be transported from lab to lab. The Filter Pack (required, sold separately) bundles all water filter technologies into one convenient twin cartridge package so installation is simple.
  • To install, the WaterPro BT requires only simple connection to a tap water supply, drain and electrical source
  • Delivers ultrapure Type I, up to 18,2 megohm-cm water at a typical rate of >0,5 liters/minute
More Product Information
Bestill nå
SPESIFIKASJONER
Modell WaterPro BT WaterPro BT (with UV lamp)
Environmentally Preferable
Tilførselsvann Tap Tap
Gjennomstrømningshastighet >0,5 l/min >0,5 l/min
TOC-innhold <10 ppb <5 ppb
B×D×H 290×422×538 mm 290×422×538 mm
Vekt 17,6 kg 18,2 kg