Cut resistant gloves, Check & Go Red Nit 1
Leverandør: HONEYWELL SAFETY
2332265/10 2332265/7 2332265/8 2332265/9 2332265/11
DALL2332265/10EA 0 NOK
DALL2332265/10 DALL2332265/7 DALL2332265/8 DALL2332265/9 112-4677
Cut resistant gloves, Check & Go Red Nit 1
Hansker
These polyamide knitted gloves feature a red oil-proof nitrile coating on palm and fingertips.
  • Knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
  • Tight knit (gauge 13) gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • Ventilated back optimises breathability
  • Nitrile coating at the palm and fingertips provides outstanding abrasion resistance and impermeability to oils
  • Colour-coding on top of the hand and coating colour helps safety manager and workers to visually recognise
More Product Information
Bestill nå
SPESIFIKASJONER
Modell Check & Go Red Nit 1
Ambidekstral/håndspesifikk Håndspesifikk
Environmentally Preferable
Engangs/flergangs gjenbrukbar
Linermateriale Polyamid
Overflate Palm and fingertips coated
Måler 13
Type mansjett Strikket håndledd
EN norm EN 388:2016 (4X21A)