Amikacin
Leverandør: MP Biomedicals
02150342.2
0215034201
0215034205
ICNA02150342.2EA
665
NOK
ICNA02150342.2
ICNA0215034201
ICNA0215034205
Amikacin
Amikacin
Sparingly soluble in water.
Formel:
C₂₂H₄₃N₅O₁₃ MW: 585,61 g/mol |
MDL nummer:
MFCD00883675 CAS-nummer: 37517-28-5 |
Spesifikasjon Testresultater
Form | White Powder |
Storage | Store at +4 °C, Store desssicated |
Loss on Drying |
|
Optical Rotation | +100,5° |
pH | 9,5 - 11,5 (1% aq soln) |
Potency | ~900 μg/mg |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...