Theophyllin, anhydrous, white powder
Leverandør: MP Biomedicals
Danger
Synonymer: 1,3-Dimethyl-1H-purine-2,6(3H,9H)-dione, Theophylline
0210302480 0210302483 0210302490 0210302491
ICNA0210302480EA 675 NOK
ICNA0210302480 ICNA0210302483 ICNA0210302490 ICNA0210302491
Theophyllin, anhydrous, white powder
Theophyllin
Phosphodiesterase inhibitor; diuretic; cardiac stimulant; muscle relaxant; asthma medication.
Soluble in 0,1 M HCl; 0,1 M NaOH; slightly soluble in water; moderately soluble in ethanol. Soluble in 1 M NH4OH (50 mg/ml - clear colorless solution) and 50% ammonium hydorxide (50 mg/ml - clear, colorless solution).
NB: Research Use Only (RUO).
Formel: C₇H₈N₄O₂
MW: 180.16 g/mol
MDL nummer: MFCD00079619
CAS-nummer: 58-55-9
FN: 2811
ADR: 6.1 III
Bestill nå
Spesifikasjon Testresultater
Identity Test Passes
FTIR Conforms to Standard